2,6-Dichlorophenol 99%

Supplier: Thermo Scientific Chemicals
Danger

11359-0010 11359-0250 11359-1000
TS11359-0010EA 500 USD
TS11359-0010 TS11359-0250 TS11359-1000
2,6-Dichlorophenol 99%
2,6-Dichlorophenol
Formula: C₆H₄Cl₂O
MW: 163 g/mol
Boiling Pt: ∼219 °C (1013 hPa)
Melting Pt: 65…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00002176
CAS Number: 87-65-0
EINECS: 201-761-3
UN: 2020
ADR: 6.1,III
Merck Index: 13,03099

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 65 to 68 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR