2,5-Dichlorophenol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11358-0010
11358-0500
11358-2500
TS11358-0010EA
522
USD
TS11358-0010
TS11358-0500
TS11358-2500
2,5-Dichlorophenol 98%
2,5-Dichlorophenol
Formula:
C₆H₄Cl₂O MW: 163 g/mol Boiling Pt: 211 °C (760 mmHg) Melting Pt: 54…58 °C Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002174 CAS Number: 583-78-8 EINECS: 209-520-4 UN: 2020 ADR: 6.1,III |
Specification Test Results
Appearance | White to light brown Crystals of flakes |
Infrared spectrum | Conforms |
Melting point | 54 to 58 °C |
GC | ≥97.5 % |
Uniformity coefficient | ≤1.60 |
Moisture holding capacity | 49.0 to 60.0 % (Cl) |
Total volume capacity | ≥1.20 eq/l (Cl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...