2,6-Dichlorobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

11320-0100 11320-0250
TS11320-0100EA 53 USD
TS11320-0100 TS11320-0250 TS11320-1000
2,6-Dichlorobenzoic acid 98+%
2,6-Dichlorobenzoic acid
Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00002418
CAS Number: 50-30-6
EINECS: 200-025-9

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 141 to 145 °C
HPLC ≥98.0 %
Titration with NaOH ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR