2,6-Di-tert-butylphenol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Di(dimethylethyl)-2-hydroxybenzene

11300-0010 11300-0025 11300-2500
TS11300-0010EA 59 USD
TS11300-0010 TS11300-0025 TS11300-2500
2,6-Di-tert-butylphenol 99%
2,6-Di-tert-butylphenol
Formula: C₁₄H₂₂O
MW: 206.33 g/mol
Boiling Pt: 253 °C (760 mmHg)
Melting Pt: 35…38 °C
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00008820
CAS Number: 128-39-2
EINECS: 204-884-0
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to light yellow Crystalline solid
Infrared spectrum Conforms
Melting point 35 to 38 °C
Water ≤0.1 % (K.F.)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR