α,α'-Dibromo-o-xylene 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Bis(bromomethyl)benzene, o-Xylylene bromide, o-Xylylene dibromide, alpha,alpha'-Dibromo-o-xylene, o-Xylene dibromide

11293-0250 11293-1000
TS11293-0250EA 81 USD
TS11293-0250 TS11293-1000
α,α'-Dibromo-o-xylene 96%
α,α'-Dibromo-o-xylene
Formula: C₈H₈Br₂
Boiling Pt: 140 °C (20 mmHg)
Melting Pt: 91…96 °C
Density: 1.96 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000175
CAS Number: 91-13-4
EINECS: 202-042-7
UN: 2928
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 91 to 96 °C
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR