trans-1,4-Diphenyl-2-butene-1,4-dione 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,2-Dibenzoylethylene, trans-1,2-Dibenzoylethylene

11258-0050 11258-0250
TS11258-0010EA 37.08 USD
TS11258-0010 TS11258-0050 TS11258-0250
trans-1,4-Diphenyl-2-butene-1,4-dione 97%
trans-1,4-Diphenyl-2-butene-1,4-dione
Formula: C₁₆H₁₂O₂
MW: 236.27 g/mol
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003083
CAS Number: 959-28-4
EINECS: 213-498-1

Order Now

Specification Test Results

Appearance Yellow to light orange Crystalline powder
Infrared spectrum Conforms
Melting point 108 to 112 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR