4-Hydroxybenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

11083-0250 11083-1000 11083-5000
TS11083-0250EA 52 USD
TS11083-0250 TS11083-1000 TS11083-5000
4-Hydroxybenzonitrile 99%
Formula: C₇H₅NO
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002312
CAS Number: 767-00-0
EINECS: 212-175-2

Order Now

Specification Test Results

Appearance Off-white to beige to light grey Crystalline powder
Infrared spectrum Conforms
Melting point 110 to 113 °C
Water ≤0.4 %
Sulfated ash ≤0.2 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR