4-Cyanobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Formylbenzonitrile

11072-0050 11072-0250 11072-1000
TS11072-0050EA 64 USD
TS11072-0050 TS11072-0250 TS11072-1000
4-Cyanobenzaldehyde 98%
4-Cyanobenzaldehyde
Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 133 °C
Melting Pt: 100…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00003376
CAS Number: 105-07-7
EINECS: 203-267-3
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 98 to 103 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR