o-Cresol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Hydroxy-2-methylbenzene, 2-Methylphenol, 2-Hydroxytoluene

11055-0010 11055-0025 11055-0100 11055-1000 11055-5000
TS11055-0010EA 163 USD
TS11055-0010 TS11055-0025 TS11055-0100 TS11055-1000 TS11055-5000
o-Cresol 99%
o-Cresol
Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 191 °C (760 mmHg)
Melting Pt: 30…32 °C
Density: 1.04 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002226
CAS Number: 95-48-7
EINECS: 202-423-8
UN: 3455
ADR: 6.1,II
Merck Index: 13,02604

Order Now

Specification Test Results

Appearance Colorless to amber Low melting solid or liquid (May darken on storage)
Infrared spectrum Conforms
Water ≤0.2 % (K.F.)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR