Cinnamyl bromide predominantly trans 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (3-Bromo-1-propen-1-yl)benzene, 3-Bromo-1-phenyl-1-propene

11040-0050 11040-0250 11040-1000
TS11040-0050EA 38 USD
TS11040-0050 TS11040-0250 TS11040-1000
Cinnamyl bromide predominantly trans 97%
Cinnamyl bromide
Formula: C₉H₉Br
MW: 197.08 g/mol
Boiling Pt: 103 °C (22 mmHg)
Melting Pt: 26…32 °C
Density: 1.332 g/cm³
Flash Pt: 87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000245
CAS Number: 4392-24-9
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance Light yellow to beige to light brown Solidified mass or fragments
Infrared spectrum Conforms
Melting point 26 to 32 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR