Cinnamoyl chloride predominantly trans 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Phenyl-2-propenoyl chloride, Cinnamic acid chloride

11039-0050 11039-1000 11039-5000
TS11039-0050EA 27 USD
TS11039-0050 TS11039-1000 TS11039-5000
Cinnamoyl chloride predominantly trans 98%
Cinnamoyl chloride
Formula: C₉H₇ClO
MW: 166.6043 g/mol
Melting Pt: 33…37 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000732
CAS Number: 102-92-1
EINECS: 203-065-5
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance White to yellow to orange Crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 33 to 37 °C
Titration RCOCl 97.5 to 102.5 %
UV at 320 nm A: ≤0.009
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity ≤0.0008 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR