trans-Cinnamonitrile predominantly trans 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (E)-3-Phenylacrylonitrile, Cinnamonitrile

11038-0050 11038-0250 11038-1000
TS11038-0050EA 29 USD
TS11038-0050 TS11038-0250 TS11038-1000
trans-Cinnamonitrile predominantly trans 97%
trans-Cinnamonitrile
Formula: C₉H₇N
MW: 129.16 g/mol
Melting Pt: 20 °C
Density: 1.02 g/cm³
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00001930
CAS Number: 1885-38-7
EINECS: 224-441-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5990 to 1.6040 (20°C, 589 nm)
GC ≤10 % (cis-Cinnamonitrile)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR