α-Chloro-p-xylene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11017-1000
11017-5000
TS11017-1000EA
52
USD
TS11017-1000
TS11017-5000
α-Chloro-p-xylene 98%
α-Chloro-p-xylene
Formula:
C₈H₉Cl Boiling Pt: 200 °C (1013 hPa) Melting Pt: 4 °C Density: 1.06 g/cm³ Flash Pt: 75 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000919 CAS Number: 104-82-5 EINECS: 203-241-1 UN: 3265 ADR: 8,II Merck Index: 12,10224 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5330 to 1.5370 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...