α-Chloro-m-xylene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-(Chloromethyl)-3-methylbenzene, 3-Methylbenzyl chloride

11016-0250 11016-1000
TS11016-0250EA 43 USD
TS11016-0250 TS11016-1000
α-Chloro-m-xylene 98%
α-Chloro-m-xylene
Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 196 °C (1013 hPa)
Density: 1.064 g/cm³
Flash Pt: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000909
CAS Number: 620-19-9
EINECS: 210-628-9
UN: 3265
ADR: 8,II
Merck Index: 12,10224

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5335 to 1.5365 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR