α-Chloro-o-xylene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methylbenzyl chloride

11015-0250 11015-1000 11015-5000
TS11015-0250EA 34 USD
TS11015-0250 TS11015-1000 TS11015-5000
α-Chloro-o-xylene 99%
α-Chloro-o-xylene
Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 197…199 °C (760 mmHg)
Melting Pt: –2 °C
Density: 1.08 g/cm³
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000899
CAS Number: 552-45-4
EINECS: 209-013-8
UN: 3265
ADR: 8,II
Merck Index: 12,10224

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5405 to 1.5425 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR