3-Chlorophenol 99%

Supplier: Thermo Scientific Chemicals
Warning

10981-0010 10981-0050 10981-0250 10981-1000
TS10981-0010EA 872 USD
TS10981-0010 TS10981-0050 TS10981-0250 TS10981-1000
3-Chlorophenol 99%
3-Chlorophenol
Formula: C₆H₅ClO
Boiling Pt: 214 °C (760 mmHg)
Melting Pt: 33…35 °C
Density: 1.218 g/cm³
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00002256
CAS Number: 108-43-0
EINECS: 203-582-6
UN: 2020
ADR: 6.1,III
Merck Index: 13,02173

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5650 to 1.5670 (20°C, 589 nm)
GC ≥98.5 %
Iron (Fe) ≤50 ppm
Magnesium (Mg) ≤100 ppm
Potassium (K) ≤50 ppm
Sodium (Na) ≤100 ppm
Silicon dioxide (SiO2) ≤1000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR