4-Chloro-3-nitrobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

10960-0050 10960-0250 10960-1000
TS10960-0050EA 45 USD
TS10960-0050 TS10960-0250 TS10960-1000
4-Chloro-3-nitrobenzaldehyde 97%
4-Chloro-3-nitrobenzaldehyde
Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Melting Pt: 61…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00007078
CAS Number: 16588-34-4
EINECS: 240-645-7

Order Now

Specification Test Results

Appearance Off-white to light yellow to light green Powder
Infrared spectrum Conforms
Melting point 60 to 65 °C
GC ≥96.0 %
Titration with HClO4 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR