4-Chloro-m-cresol 99+%

Supplier: Thermo Scientific Chemicals
Danger

10948-0050 10948-1000 10948-5000
TS10948-0050EA 25 USD
TS10948-0050 TS10948-1000 TS10948-5000
4-Chloro-m-cresol 99+%
4-Chloro-m-cresol
Formula: C₇H₇ClO
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 63…65 °C
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00002323
CAS Number: 59-50-7
EINECS: 200-431-6
UN: 3261
ADR: 8,III
Merck Index: 13,02152

Order Now

Specification Test Results

Appearance White to almost white Crystals, flakes, pellets or crystalline powder
Infrared spectrum Conforms
Melting point 64 to 67 °C
GC ≥99.0 %
Density (20 °C) 0.702 to 0.715 g/ml
Additional info Bromine index < 100 ppm
Color scale ≤10 APHA
Benzene ≤5 ppm
Aromatics ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR