5-Chloro-o-toluidine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Chloro-2-methylaniline, 2-Amino-4-chlorotoluene
10938-0050
10938-1000
10938-5000
TS10938-0050EA
33
USD
TS10938-0050
TS10938-1000
TS10938-5000
5-Chloro-o-toluidine 98%
5-Chloro-o-toluidine
Formula:
C₇H₈ClN Boiling Pt: 237 °C (1013 hPa) Melting Pt: 22 °C Flash Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007779 CAS Number: 95-79-4 EINECS: 202-452-6 UN: 2239 ADR: 6.1,III |
Specification Test Results
Appearance | Clear yellow to light brown Liquid after melting (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5830 to 1.5850 (20°C, 589 nm) |
GC | ≥97.5 % |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...