4-Chloro-o-toluidine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Amino-5-chlorotoluene, 4-Chloro-2-methylaniline, 4-Chloro-o-toluidine, Fast Red TR Base

10937-0050 10937-1000
TS10937-0050EA 36 USD
TS10937-0050 TS10937-1000
4-Chloro-o-toluidine 98%
4-Chloro-o-toluidine
Formula: C₇H₈ClN
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 29…30 °C
Density: 1.19 g/cm³
Flash Pt: 124 °C
MDL Number: MFCD00007842
CAS Number: 95-69-2
EINECS: 202-441-6
UN: 2239
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5845 to 1.5865 (20°C, 589 nm)
GC ≥97.5 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR