4-Chloro-3,5-xylenol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-5-hydroxy-3-xylene, Chloroxylenol, 4-Chloro-3,5-dimethylphenol
10906-0025
10906-1000
10906-5000
TS10906-0025EA
296
USD
TS10906-0025
TS10906-1000
TS10906-5000
4-Chloro-3,5-xylenol 99%
4-Chloro-3,5-xylenol
Formula:
C₈H₉ClO Boiling Pt: 246 °C (760 mmHg) Melting Pt: 113…116 °C Flash Pt: 132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002324 CAS Number: 88-04-0 EINECS: 201-793-8 UN: 3077 ADR: 9,III Merck Index: 13,02194 |
Specification Test Results
Appearance | White to light beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 113 to 116 °C |
GC | ≥98.5 % |
Stabilizer | ca. 500 ppm MEHQ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...