(4-Chlorophenyl)acetonitrile 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Chlorobenzyl cyanide

10881-0050 10881-1000
TS10881-0050EA 26 USD
TS10881-0050 TS10881-1000 TS10881-2500 TS10881-5000
(4-Chlorophenyl)acetonitrile 98+%
(4-Chlorophenyl)acetonitrile
Formula: C₈H₆ClN
MW: 151.6 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 29…30 °C
Flash Pt: 133 °C
Storage Temperature: Ambient
MDL Number: MFCD00001918
CAS Number: 140-53-4
EINECS: 205-418-9
UN: 3439
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5415 to 1.5435 (20°C, 589 nm)
Water ≤0.3 % (K.F.)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR