2-Chlorobenzylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (2-Chlorophenyl)methanamine

10878-0250 10878-1000
TS10878-0250EA 45 USD
TS10878-0250 TS10878-1000
2-Chlorobenzylamine 97%
2-Chlorobenzylamine
Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 103…104 °C (11 mmHg)
Density: 1.17 g/cm³
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00008108
CAS Number: 89-97-4
EINECS: 201-955-8
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5610 to 1.5640 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR