3-Chlorobenzoyl chloride 99+%

Supplier: Thermo Scientific Chemicals
Danger

10874-0050 10874-0500 10874-1000 10874-5000
TS10874-0050EA 28 USD
TS10874-0050 TS10874-0500 TS10874-1000 TS10874-5000
3-Chlorobenzoyl chloride 99+%
3-Chlorobenzoyl chloride
Formula: C₇H₄Cl₂O
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: 44 °C
Density: 1.36 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000671
CAS Number: 618-46-2
EINECS: 210-552-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5680 to 1.5700 (20°C, 589 nm)
GC ≤0.03 % Total other poly-halogenated acid halides
Titration RCOCl ≥99.0 %
Titration RCOOH ≤1.0 % (as Chlorobenzoic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR