3-Chlorobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: m-Chlorobenzonitrile

10868-0100 10868-0500
TS10868-0100EA 58 USD
TS10868-0100 TS10868-0500
3-Chlorobenzonitrile 99%
3-Chlorobenzonitrile
Formula: C₇H₄ClN
MW: 137.57 g/mol
Boiling Pt: 203
Melting Pt: 39
Storage Temperature: Ambient
MDL Number: MFCD00001798
CAS Number: 766-84-7
EINECS: 212-172-6

Order Now

Specification Test Results

Appearance Off-white to light beige Crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 38 to 42 °C
GC ≥98.5 %
Stabilizer 50 to 150 ppm Amylene


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR