2-Chlorobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

10867-0010 10867-2500
TS10867-0010EA 200 USD
TS10867-0010 TS10867-2500
2-Chlorobenzonitrile 99%
2-Chlorobenzonitrile
Formula: C₇H₄ClN
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 43…46 °C
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00001779
CAS Number: 873-32-5
EINECS: 212-836-5

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals and/or chunks or solid
Infrared spectrum Conforms
Melting point 43 to 46 °C
Water ≤0.5 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR