3-Chlorobenzoic acid 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
10866-0250
10866-1000
10866-5000
TS10866-0250EA
33
USD
TS10866-0250
TS10866-1000
TS10866-5000
3-Chlorobenzoic acid 99+%
3-Chlorobenzoic acid
Formula:
C₇H₅ClO₂ MW: 156.57 g/mol Melting Pt: 154…157 °C Flash Pt: 150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002491 CAS Number: 535-80-8 EINECS: 208-618-4 Merck Index: 13,02142 |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 154 to 157 °C |
HPLC | ≥99.0 % |
Titration with NaOH | 99.0 to 100.5 % |
BSC Certification | Passes |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...