4-Chloroaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

10859-0010 10859-0050 10859-1000 10859-5000
TS10859-0010EA 118 USD
TS10859-0010 TS10859-0050 TS10859-1000 TS10859-5000
4-Chloroaniline 98%
4-Chloroaniline
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 232 °C (760 mmHg)
Melting Pt: 68…72 °C
Flash Pt: >188 °C
Storage Temperature: Ambient
MDL Number: MFCD00007835
CAS Number: 106-47-8
EINECS: 203-401-0
UN: 2018
ADR: 6.1,II
Merck Index: 13,02136

Order Now

Specification Test Results

Appearance Beige to brown to purple Crystalline solid (May darken on storage)
Infrared spectrum Conforms
Melting point 68 to 72 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR