2-tert-Butyl-p-cresol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-tert-Butyl-2-hydroxy-5-methylbenzene, 2-tert-Butyl-4-methylphenol, 3-tert-Butyl-4-hydroxytoluene

10796-0050 10796-1000 10796-5000
TS10796-0050EA 28 USD
TS10796-0050 TS10796-1000 TS10796-5000
2-tert-Butyl-p-cresol 99%
2-tert-Butyl-p-cresol
Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 50…53 °C
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00002381
CAS Number: 2409-55-4
EINECS: 219-314-6
UN: 2430
ADR: 8,III

Order Now

Specification Test Results

Appearance White to yellow to light beige Crystalline mass
Infrared spectrum Conforms
Melting point 50 to 53 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR