2,3-Butanediol mixture of racemic and meso forms 98% tech.
Supplier: Thermo Scientific Chemicals
Synonyms:
2,3-Dihydroxy butane, (±)-2,3-Butanediol, 2,3-Butylene glykol
10764-0010
10764-0025
10764-2500
TS10764-0010EA
760
USD
TS10764-0010
TS10764-0025
TS10764-2500
2,3-Butanediol mixture of racemic and meso forms 98% tech.
2,3-Butanediol
Formula:
C₄H₁₀O₂ MW: 90.12 g/mol Boiling Pt: 180 °C (760 mmHg) Melting Pt: 19 °C Density: 1.01 g/cm³ Flash Pt: 85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004523 CAS Number: 513-85-9 EINECS: 208-173-6 Merck Index: 12,01602 |
Specification Test Results
Appearance | Clear colorless to light yellow Viscous liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4330 to 1.4370 (20°C, 589 nm) |
GC | ≥97.5 % |
Absorption Ratio P-15/P+15 | 0.98 to 1.20 |
BSC Certification | Passes |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...