α-Bromo-p-xylene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: alpha-Bromo-p-xylene, 4-Methylbenzyl bromide, 4-(Bromomethyl)toluene

10757-0050 10757-0250 10757-1000
TS10757-0050EA 27 USD
TS10757-0050 TS10757-0250 TS10757-1000
α-Bromo-p-xylene 98%
α-Bromo-p-xylene
Formula: C₈H₉Br
Boiling Pt: 218…220 °C (760 mmHg)
Melting Pt: 33…37 °C
Flash Pt: 97 °C
Storage Temperature: Ambient
MDL Number: MFCD00000181
CAS Number: 104-81-4
EINECS: 203-240-6
UN: 3417
ADR: 6.1,II
Merck Index: 12,10223

Order Now

Specification Test Results

Appearance White to light yellow Crystalline mass
Infrared spectrum Conforms
Melting point 33 to 37 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR