4-Bromoveratrole 97%

Supplier: Thermo Scientific Chemicals
Warning

10754-0050 10754-0250 10754-1000
TS10754-0050EA 20 USD
TS10754-0050 TS10754-0250 TS10754-1000
4-Bromoveratrole 97%
4-Bromoveratrole
Formula: C₈H₉BrO₂
Boiling Pt: 255 °C (1013 hPa)
Density: 1.5 g/cm³
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00008381
CAS Number: 2859-78-1
EINECS: 220-677-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5705 to 1.5755 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR