4-Bromotoluene 99%

Supplier: Thermo Scientific Chemicals
Warning

10748-1000 10748-5000
TS10748-1000EA 69 USD
TS10748-1000 TS10748-5000
4-Bromotoluene 99%
4-Bromotoluene
Formula: C₇H₇Br
Boiling Pt: 184 °C (1013 hPa)
Melting Pt: 26…29 °C
Density: 1.39 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00000109
CAS Number: 106-38-7
EINECS: 203-391-8
Merck Index: 13,01424

Order Now

Specification Test Results

Appearance White Low melting crystalline mass
Infrared spectrum Conforms
GC ≥99.0 %
Water ≤0.05 %
Color scale ≤10 APHA
Stabilizer 10 to 20 ppm MEHQ
Acidity ≤0.01 % (as acrylic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR