2-Bromotoluene 99%

Supplier: Thermo Scientific Chemicals
Warning

10747-0250 10747-1000
TS10747-0250EA 52 USD
TS10747-0250 TS10747-1000
2-Bromotoluene 99%
2-Bromotoluene
Formula: C₇H₇Br
Boiling Pt: 181.5 °C (1013 hPa)
Melting Pt: –27 °C
Density: 1.42 g/cm³
Flash Pt: 78 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000068
CAS Number: 95-46-5
EINECS: 202-421-7
Merck Index: 13,01424

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5550 to 1.5570 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR