4-Bromoresorcinol 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Bromobenzene-1,3-diol

10743-0050 10743-0250
TS10743-0050EA 109 USD
TS10743-0050 TS10743-0250
4-Bromoresorcinol 98%
4-Bromoresorcinol
Formula: C₆H₅BrO₂
MW: 189.01 g/mol
Boiling Pt: 150 °C (12 mmHg)
Melting Pt: 99…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00002272
CAS Number: 6626-15-9
EINECS: 229-586-8

Order Now

Specification Test Results

Appearance Beige to pink Powder or crystals
Infrared spectrum Conforms
Melting point 99 to 104 °C
GC ≥99.0 %
Color scale ≤10 APHA
Stabilizer ≤10 ppm BHT
Residue after evaporation ≤0.001 %
Carbonyl (as HCHO) ≤0.001 %
Titrable acid ≤0.0002 meq/g
E/E (limit about 0.05%) Passes test
Peroxides ≤1 ppm (as H2O2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR