6-Bromo-2-naphthol 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Bromo-2-hydroxynaphthalene, 2-Bromo-6-hydroxynaphthalene
10717-0250
10717-1000
TS10717-0250EA
71
USD
TS10717-0250
TS10717-1000
6-Bromo-2-naphthol 97%
6-Bromo-2-naphthol
Formula:
C₁₀H₇BrO Melting Pt: 125…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004081 CAS Number: 15231-91-1 EINECS: 239-279-0 |
Specification Test Results
Appearance | Off-white to beige Powder |
Infrared spectrum | Conforms |
Melting point | 125 to 130 °C |
GC | ≥96.0 % |
Color of solution | ≤100 APHA (10% aq. soln.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...