1-Bromo-2-naphthol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromonaphthalen-2-ol, 1-Bromo-2-hydroxynaphthalene

10716-0250 10716-1000
TS10716-0250EA 61 USD
TS10716-0250 TS10716-1000
1-Bromo-2-naphthol 98%
1-Bromo-2-naphthol
Formula: C₁₀H₇BrO
MW: 223.07 g/mol
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 77…81 °C
MDL Number: MFCD00003869
CAS Number: 573-97-7
EINECS: 209-363-1

Order Now

Specification Test Results

Appearance Off-white to beige to brown-purple Crystalline powder
Infrared spectrum Conforms
Melting point 77 to 84 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR