1-Bromo-4-fluorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
10701-0100
10701-1000
10701-5000
TS10701-0100EA
30
USD
TS10701-0100
TS10701-1000
TS10701-5000
1-Bromo-4-fluorobenzene 99%
1-Bromo-4-fluorobenzene
Formula:
C₆H₄BrF Boiling Pt: 151 °C (1013 hPa) Melting Pt: –8 °C Density: 1.593 g/cm³ Flash Pt: 53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000342 CAS Number: 460-00-4 EINECS: 207-300-2 UN: 1992 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5260 to 1.5280 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...