1-Bromo-3-fluorobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromofluorobenzene

10700-0250 10700-1000
TS10700-0250EA 50 USD
TS10700-0250 TS10700-1000
1-Bromo-3-fluorobenzene 99%
1-Bromo-3-fluorobenzene
Formula: C₆H₄BrF
MW: 175 g/mol
Boiling Pt: 150 °C (760 mmHg)
Melting Pt: –8 °C
Density: 1.567 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00000326
CAS Number: 1073-06-9
EINECS: 214-023-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5260 to 1.5280 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR