4-Bromo-3,5-xylenol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-3,5-dimethylphenol, 2-Bromo-5-hydroxy-3-xylene

10687-0100 10687-0250 10687-1000 10687-5000
TS10687-0100EA 91 USD
TS10687-0100 TS10687-0250 TS10687-1000 TS10687-5000
4-Bromo-3,5-xylenol 99%
4-Bromo-3,5-xylenol
Formula: C₈H₉BrO
Melting Pt: 113…116 °C
Storage Temperature: Ambient
MDL Number: MFCD00002315
CAS Number: 7463-51-6
EINECS: 231-255-8

Order Now

Specification Test Results

Appearance Colorless to light pink Powder or crystals
Infrared spectrum Conforms
Melting point 113 to 116 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR