1-Bromo-4-chlorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
10683-0010
10683-0500
10683-1000
10683-5000
TS10683-0010EA
100
USD
TS10683-0010
TS10683-0500
TS10683-1000
TS10683-5000
1-Bromo-4-chlorobenzene 99%
1-Bromo-4-chlorobenzene
Formula:
C₆H₄BrCl Boiling Pt: 196 °C (760 mmHg) Melting Pt: 64…67 °C Density: 1.651 g/cm³ Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000600 CAS Number: 106-39-8 EINECS: 203-392-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 64 to 67 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...