2-Bromobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromobenzene-1-carbonyl chloride, o-Bromobenzoyl chloride

10675-0100 10675-0500
TS10675-0100EA 60 USD
TS10675-0100 TS10675-0500
2-Bromobenzoyl chloride 98%
2-Bromobenzoyl chloride
Formula: C₇H₄BrClO
MW: 219.46 g/mol
Boiling Pt: 245
Melting Pt: 11
Density: 1.67 g/cm³
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000655
CAS Number: 7154-66-7
EINECS: 230-507-4
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5947 to 1.5967 (20°C, 589 nm)
Titration RCOCl ≥97.5 %
UV at 420 nm A: ≤0.03
Color scale ≤15 APHA
Residue after evaporation ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR