3-Bromobenzaldehyde 96%

Supplier: Thermo Scientific Chemicals
Warning

10666-0250 10666-1000
TS10666-0250EA 37 USD
TS10666-0250 TS10666-1000
3-Bromobenzaldehyde 96%
3-Bromobenzaldehyde
Formula: C₇H₅BrO
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: 18…21 °C
Density: 1.58 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00003345
CAS Number: 3132-99-8
EINECS: 221-526-9

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5920 to 1.5960 (20°C, 589 nm)
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR