2-Bromobenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

10665-0100 10665-0250 10665-1000
TS10665-0100EA 35 USD
TS10665-0100 TS10665-0250 TS10665-1000
2-Bromobenzaldehyde 97%
2-Bromobenzaldehyde
Formula: C₇H₅BrO
Boiling Pt: 230 °C (760 mmHg)
Melting Pt: 21…22 °C
Density: 1.58 g/cm³
Flash Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003300
CAS Number: 6630-33-7
EINECS: 229-622-2

Order Now

Specification Test Results

Appearance Clear yellow Liquid after melting
Infrared spectrum Conforms
Refractive index 1.5950 to 1.5970 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR