9-Bromoanthracene 96%

Supplier: Thermo Scientific Chemicals
Warning

10664-0050 10664-0250
TS10664-0050EA 64 USD
TS10664-0050 TS10664-0250
9-Bromoanthracene 96%
9-Bromoanthracene
Formula: C₁₄H₉Br
MW: 257.13 g/mol
Boiling Pt: 359 °C (1013 hPa)
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00001243
CAS Number: 1564-64-3
EINECS: 216-359-3

Order Now

Specification Test Results

Appearance Yellow to green to brown Powder or crystals or needles
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR