3-Bromoanisole 99+%

Supplier: Thermo Scientific Chemicals

10662-0250 10662-1000
TS10662-0250EA 41 USD
TS10662-0250 TS10662-1000
3-Bromoanisole 99+%
3-Bromoanisole
Formula: C₇H₇BrO
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.47 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00000081
CAS Number: 2398-37-0
EINECS: 219-264-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5630 to 1.5650 (20°C, 589 nm)
Water ≤0.1 %
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR