2-Bromoaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
10661-0250
10661-1000
10661-5000
TS10661-5000EA
800
USD
TS10661-5000
TS10661-0250
TS10661-1000
2-Bromoaniline 98%
2-Bromoaniline
Formula:
C₆H₆BrN MW: 172.02 g/mol Boiling Pt: 229 °C (760 mmHg) Melting Pt: 29…31 °C Density: 1.578 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007632 CAS Number: 615-36-1 EINECS: 210-421-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Clear yellow to red-brown Liquid after melting |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Titration with HClO4 | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...