Benzyltrimethylammonium hydroxide 40% (w/w) in methanol
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Triton B, Benzyltrimethylammonium Hydroxide, N,N,N-Trimethyl-1-phenylmethanaminium Hydroxide
10620-0010
10620-0051
10620-2500
TS10620-0010EA
207
USD
TS10620-0010
TS10620-0051
TS10620-2500
Benzyltrimethylammonium hydroxide 40% (w/w) in methanol
Benzyltrimethylammonium hydroxide
Formula:
C₁₀H₁₇NO MW: 167.25 g/mol Density: 0.925 g/cm³ Flash Pt: 15 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008281 CAS Number: 100-85-6 EINECS: 202-895-5 UN: 3286 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to light yellow Solution |
Infrared spectrum | Conforms |
Titration with HCl | ≥40 % |
Titration with HClO4 | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...