Benzyl(methyl)amine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Benzylmethylamine, N-Methyl-1-phenylmethanamine, N-Methylbenzylamine

10604-0050 10604-5000
TS10604-0050EA 22 USD
TS10604-0050 TS10604-5000
Benzyl(methyl)amine 97%
Benzyl(methyl)amine
Formula: C₆H₅CH₂NHCH₃
MW: 121.18 g/mol
Boiling Pt: ∼184 °C (1013 hPa)
Melting Pt: –24 °C
Density: 0.94 g/cm³
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00008289
CAS Number: 103-67-3
EINECS: 203-133-4
UN: 2922
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=067337f4cbd3ebd20fe522a5bac5c9f29cb10e6a&vid=fab9de39bcd5ba3758c9402492897f6aed55e8ee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5214 to 1.5234 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥96.0 %
Titration with HClO4 ≥96.0 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Titrable acid ≤0.0003 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR