Phenylglyoxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Oxo-2-phenylacetic acid, Benzoylformic acid

10576-0050 10576-0250 10576-1000
TS10576-0050EA 44 USD
TS10576-0050 TS10576-0250 TS10576-1000
Phenylglyoxylic acid 97%
Phenylglyoxylic acid
Formula: C₈H₆O₃
MW: 150.13 g/mol
Melting Pt: 60…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00002575
CAS Number: 611-73-4
EINECS: 210-278-7

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 60 to 69 °C
Titration with NaOH ≥96.0 %
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR